Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
        Novus Biologicals™ Mitochondria Isolation Kit
    
    
    
        
            Shop All R&D Systems Products
        
    
    
    
    
    
    
    
    
        
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                1 Kit
                            
                        
                    Specifications
Specifications
| Content And Storage | Storage is content dependent. | 
| Quantity | 1 Kit | 
| Kit Contents | 5X Homogenization Buffer 50mL, 5X Suspension Buffer 2 x 50mL, Mitochondrial Staining Solution 1mL, Mitochondria Lysis Buffer 50mL, 10X PBS 2 x 50mL, 1M DTT 100μL, 100X Protease Inhibitor Cocktail (PIC) 100μL | 
| For Use With (Application) | Western Blotting | 
| Product Type | Mitochondrial Extraction Kit | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction