Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse 4-1BB Ligand/TNFSF9 Protein
SDP

Catalog No. nbp226580 Shop All Novus Biologicals Products
Click to view available options
:
0.05mg; Unlabeled

Highly purified. Generating reliable and reproducible results. Applications: Functional, SDS-Page

A recombinant protein corresponding to TNFSF9. The Recombinant Mouse 4-1BB Ligand/TNFSF9 Protein has been validated for the following applications: Functional, SDS-Page.

Specifications

Gene ID (Entrez) 8744
Name 4-1BB Ligand/TNFSF9 protein
Purification Method Protein A purified
Storage Requirements −20°C. Avoid freeze-thaw cycles.
Cross Reactivity Mouse
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.