Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ NCF1 Protein
SDP

Catalog No. p-5355068 Shop All R&D Systems Products
Click to view available options
Quantity:
0.05 mg
0.25 mg

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-390 of Human NCF1 The Recombinant Human NCF1 Protein is derived from E. coli. The Recombinant Human NCF1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number AAH02816
Concentration 0.5mg/mL
For Use With (Application) ELISA, SDS-PAGE
Formulation In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 20% glycerol, 0.1M NaCl
Gene ID (Entrez) 653361
Molecular Weight (g/mol) 46kDa
Purification Method Protein
Quantity 0.05 mg
Source E. coli
Immunogen NCF1, 1-390aa, Human, His tag, E. coli (AAH02816).
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name NCF1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.