Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ NCK1 Protein
Shop All Novus Biologicals Products

Click to view available options
Quantity:
0.1 mg
0.5 mg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-377 of Human NCK1 The Recombinant Human NCK1 Protein is derived from E. coli. The Recombinant Human NCK1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Concentration | 1.0mg/mL |
For Use With (Application) | ELISA, SDS-PAGE |
Formulation | In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 20% glycerol, 1mM EDTA, 50mM NaCl |
Gene ID (Entrez) | 4690 |
Molecular Weight (g/mol) | 45kDa |
Purification Method | Protein |
Quantity | 0.1 mg |
Source | Human |
Immunogen | MGSSHHHHHH SSGLVPRGSH MAEEVVVVAK FDYVAQQEQE LDIKKNERLW LLDDSKSWWR VRNSMNKTGF VPSNYVERKN SARKASIVKN LKDTLGIGKV KRKPSVPDSA SPADDSFVDP GERLYDLNMP AYVKFNYMAE REDELSLIKG TKVIVMEKCS DGWWRGSYNG QVGWFPSNYV TEEGDSPLGD HVGSLSEKLA AVVNNLNTGQ VLHVVQALYP FSSSNDEELN FEKGDVMDVIEKPENDPEWW KCRKINGMVG LVPKNYVTVM QNNPLTSGLE PSPPQCDYIR PSLTGKFAGN PWYYGKVTRH QAEMALNERG HEGDFLIRDS ESSPNDFSVS LKAQGKNKHF KVQLKETVYC IGQRKFSTME ELVEHYKKAP IFTSEQGEKL YLVKHLS |
Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction