Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ PKM2 Antibody Blocking Peptide
Shop All Novus Biologicals Products

Click to view available options
Quantity:
0.05 mL
Specifications
Specifications
Gene ID (Entrez) | 5315 |
Species | Mouse |
Accession Number | P14618 |
Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
Formulation | Peptide dissolved in dH2O. Contains no BSA. |
For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-48308. For Further Blocking Peptide Related Protocol, Click Here . |
Gene Alias | CTHBP, Cytosolic thyroid hormone-binding protein, MGC3932, OIP-3, OIP3EC 2.7.1.40, Opa-interacting protein 3, Pk., muscle type, PK2, PK3PKM, Pyruvate kinase 2/3, pyruvate kinase isozymes M1/M2, Pyruvate kinase muscle isozyme, pyruvate kinase, muscle, TCB, THBP1p58, Thyroid hormone-binding protein 1, thyroid hormone-binding protein, cytosolic, Tumor M2-Pk. |
Gene Symbol | PKM |
Product Type | PKM2 |
Quantity | 0.05 mL |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction