Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ RhoD Protein
Shop All R&D Systems Products

Click to view available options
Quantity:
0.01 mg
0.05 mg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 18-207 of Human RhoD The Recombinant Human RhoD Protein is derived from E. coli. The Recombinant Human RhoD Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Concentration | 0.25mg/mL |
For Use With (Application) | SDS-PAGE |
Formulation | Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 5mM DTT, 50% glycerol, 200mM NaCl, 2mM EDTA, 0.1mM PMSF |
Molecular Weight (g/mol) | 23.8kDa |
Purification Method | SDS-PAGE |
Quantity | 0.01 mg |
Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
Regulatory Status | RUO |
Common Name | RhoD |
Conjugate | Unconjugated |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction