Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ SCP1 Antibody Blocking Peptide
Shop All Novus Biologicals Products

Click to view available options
Quantity:
0.05 mg
Description
A peptide to SYCP1.
Specifications
Specifications
Gene ID (Entrez) | 6847 |
Species | Human |
Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
Formulation | Peptide dissolved in dH2O. Contains no BSA. |
For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB 300-228. For Further Blocking Peptide Related Protocol, Click Here . |
Gene Alias | Cancer/testis antigen 8, CT8MGC104417, HOM-TES-14, SCP1SCP-1, synaptonemal complex protein 1 |
Gene Symbol | SYCP1 |
Product Type | SCP1 |
Quantity | 0.05 mg |
Regulatory Status | RUO - research use only |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction