Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Thyroid Tissue Slides (Chronic Thyroiditis)- Paraffin
Shop All Novus Biologicals Products

Click to view available options
Species:
Human
Description
These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 60 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.
Specifications
Specifications
Cross Reactivity | Human |
Type | Thyroid Tissue Slides (Chronic Thyroiditis) |
Regulatory Status | This product is for research use only and is not approved for use in humans or in clinical diagnosis. |
Species | Human |
Tissue Condition | Chronic Thyroiditis |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction