Learn More
Description
Specifications
Specifications
| For Use With (Application) | Western Blot |
| Cell Line | Human TLR1 transfected cell lysate |
| Storage Buffer | 1X SDS sample buffer: 60mM Tris-Hcl (pH 6.8) 10mM EDTA 2 % SDS 0.03% Bromophenol Blue 10% glycerol 0.3% Beta-mercaptoethanol |
| Host Species | Human |
| Content And Storage | -80°C in the dark. Avoid freeze-thaw cycles. |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
