Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Translin Protein
SDP

Catalog No. NBP14944901 Shop All R&D Systems Products
Click to view available options
Quantity:
0.1 mg
0.5 mg

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-228 of Human Translin The Recombinant Human Translin Protein is derived from E. coli. The Recombinant Human Translin Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1mg/mL
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 10% glycerol. 100mM NaCl
Gene ID (Entrez) 7247
Molecular Weight (g/mol) 26.1kDa
Purification Method Protein
Quantity 0.1 mg
Source Human
Immunogen TSN, 1- 228 aa. Sequence: MSVSEIFVELQGFLAAEQDIREEIRKVVQSLEQTAREILTLLQGVHQGAGFQDIPKRCLKAREHFGTVKTHLTSLKTKFPAEQYYRFHEHWRFVLQRLVFLAAFVVYLETETLVTREAVTEILGIEPDREKGFHLDVEDYLSGVLILASELSRLSVNSVTAGDYSRPLHISTFINELDSGFRLLNLKNDSLRKRYDGLKYDVKKVEEVVYDLSIRGFNKETAAACVEK
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name Translin
Conjugate Unconjugated
Purity or Quality Grade >90%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.