Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ WIBG Protein
Shop All R&D Systems Products

Click to view available options
Quantity:
0.05 mg
0.25 mg
Description
A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-204 of Human WIBG The Recombinant Human WIBG Protein is derived from E. coli. The Recombinant Human WIBG Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 0.5mg/mL |
| For Use With (Application) | ELISA, Western Blot |
| Formulation | 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol 0.1M NaCl |
| Gene ID (Entrez) | 84305 |
| Molecular Weight (g/mol) | 23.7kDa |
| Purification Method | Protein |
| Quantity | 0.05 mg |
| Source | Human |
| Immunogen | MEAAGSPAAT ETGKYIASTQ RPDGTWRKQR RVKEGYVPQE EVPVYENKYV KFFKSKPELP PGLSPEATAP VTPSRPEGGE PGLSKTAKRN LKRKEKRRQQ QEKGEAEALS RTLDKVSLEE TAQLPSAPQG SRAAPTAASD QPDSAATTEK AKKIKNLKKK LRQVEELQQR IQAGEVSQPS KEQLEKLARR RALEEELEDL ELGLLEHHHH HH |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction