Learn More
Novus Biologicals™ PiBind Resin
SDP
Catalog No. 5010015k Shop All R&D Systems Products
Click to view available options
Quantity:
5000 mg
PiBind™ resin provides quick and easy way to remove contaminating Pi from buffers
PiBind resin has an extremely high affinity for inorganic phosphate, and is used to eliminate contaminating Pi from buffer solutions to reduce background signals in phosphatase or ATPase assays. PiBind resin may also be used to remove Pi from cell and tissue extracts and is a rapid alternative to desalting or dialysis. The resin works over a broad range of pH values and is unaffected by many commonly used buffer additives.
Recommended Storage
Store at 4°C. Do not freeze.
Specifications
Type Resin
Quantity 5000 mg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.