Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOX1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB396419 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396419 0.025 mg
NBP241290 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB396419 Supplier Novus Biologicals Supplier No. NBP2412900.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NOX1 Polyclonal specifically detects NOX1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.

Specifications

Antigen NOX1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Alias EC 1.6.3, GP91-2, Mitogenic oxidase 1, MOX-1, MOX1NADH/NADPH mitogenic oxidase subunit P65-MOX, NADPH oxidase 1, NADPH oxidase homolog-1, NOH1mitogenic oxidase (pyridine nucleotide-dependent superoxide-generating), NOH-1NADPH oxidase 1 variant NOH-1L, NOX-1
Gene Symbols NOX1
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid peptide near the center of human NOX1. The immunogen is located within amino acids 210 - 260 of NOX1. Amino Acid Squence: RGQTEESMNESHPRK
Molecular Weight of Antigen 64 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27035
Test Specificity NOX1 antibody is human specific.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.