Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOX1 Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB201220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB201220 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB201220 Supplier Novus Biologicals Supplier No. NBP241290JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NOX1 Polyclonal antibody specifically detects NOX1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen NOX1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias EC 1.6.3, GP91-2, Mitogenic oxidase 1, MOX-1, MOX1NADH/NADPH mitogenic oxidase subunit P65-MOX, NADPH oxidase 1, NADPH oxidase homolog-1, NOH1mitogenic oxidase (pyridine nucleotide-dependent superoxide-generating), NOH-1NADPH oxidase 1 variant NOH-1L, NOX-1
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid peptide near the center of human NOX1. The immunogen is located within amino acids 210 - 260 of NOX1. Amino Acid Squence: RGQTEESMNESHPRK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Direct Reversal of DNA Damage, Endocrinology, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 27035
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.