Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nox4 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB354913 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB354913 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB354913 Supplier Novus Biologicals Supplier No. NB11058849MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Nox4 Polyclonal antibody specifically detects Nox4 in Human, Mouse, Rat, Porcine, Bovine, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, KnockDown

Specifications

Antigen Nox4
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, KnockDown
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Knockdown Validated
Formulation 50mM Sodium Borate
Gene Alias EC 1.6.3, Kidney oxidase-1, Kidney superoxide-producing NADPH oxidase, KOX, KOX-1Renal NAD(P)H-oxidase, NADPH oxidase 4, RENOXEC 1.6.3.-
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human NOX4 protein (between residues 100-200) [UniProt Q9NPH5].
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Diabetes Research, Hypoxia, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 50507
Target Species Human, Mouse, Rat, Pig, Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.