Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Noxa Antibody, Novus Biologicals™
SDP

Catalog No. NB10056368T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056368T 0.025 mg
NB10056368 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056368T Supplier Novus Biologicals Supplier No. NB10056368SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 5 publications

Noxa Polyclonal specifically detects Noxa in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Noxa
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 2 ug/mL, Immunohistochemistry 1:50 - 1:250, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:50 - 1:250. Use reported in scientific literature (Inta et al. (2006))
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q9JM54
Gene Alias adult T cell leukemia-derived PMA-responsive, APR, Immediate-early-response protein APR, NOXAPMA-induced protein 1, phorbol-12-myristate-13-acetate-induced protein 1, PMA-Induced Protein 1, Protein Noxa
Gene Symbols PMAIP1
Host Species Rabbit
Immunogen This Noxa antibody was developed by immunizing rabbits with a mixture of synthetic peptides containing amino acids 51-66 and 75-90 of mouse Noxa.
Molecular Weight of Antigen 11.6 kDa
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5366
Test Specificity Amino acids 51-66 of mouse Noxa are 100% homologous in mouse and 77% in rat; amino acids 75-90 of mouse Noxa are 100% homologous in mouse, 93% homologous in rat and 76% homologous in human.
Target Species Human, Mouse, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.