Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Noxa Antibody (69N15C9), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP225266V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP225266V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP225266V Supplier Novus Biologicals Supplier No. NBP225266V
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Noxa Monoclonal specifically detects Noxa in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Noxa
Applications Western Blot
Classification Monoclonal
Clone 69N15C9
Conjugate DyLight 405
Dilution Western Blot
Gene Alias adult T cell leukemia-derived PMA-responsive, APR, Immediate-early-response protein APR, NOXAPMA-induced protein 1, phorbol-12-myristate-13-acetate-induced protein 1, PMA-Induced Protein 1, Protein Noxa
Gene Symbols PMAIP1
Host Species Rat
Immunogen This Noxa antibody was developed by immunizing mice with a fusion protein containing human Noxa.
Molecular Weight of Antigen 11.6 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cellular Markers, Membrane Trafficking and Chaperones, Plasma Membrane Markers, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5366
Test Specificity This product reacts with Mouse.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.