Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Noxa Antibody (69N15C9), Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP225251SS
Description
Noxa Monoclonal specifically detects Noxa in Mouse samples. It is validated for Western Blot.Specifications
Noxa | |
Monoclonal | |
0.5 mg/mL | |
Western Blot 5 - 10 ug/mL | |
adult T cell leukemia-derived PMA-responsive, APR, Immediate-early-response protein APR, NOXAPMA-induced protein 1, phorbol-12-myristate-13-acetate-induced protein 1, PMA-Induced Protein 1, Protein Noxa | |
Rat | |
11.6 kDa | |
0.025 mg | |
Apoptosis, Cancer, Cellular Markers, Membrane Trafficking and Chaperones, Plasma Membrane Markers, Stem Cell Markers, Tumor Suppressors | |
5366 | |
Mouse | |
Purified |
Western Blot | |
69N15C9 | |
Unconjugated | |
NP_067426.1 | |
PMAIP1 | |
This Noxa antibody (69N15C9) is made against amino acids 1-103 of mouse Noxa. | |
Protein G purified | |
RUO | |
Primary | |
This product reacts with Mouse. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction