Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Noxa Antibody (69N15C9), Novus Biologicals™
SDP

Rat Monoclonal Antibody

$181.90 - $402.00

Specifications

Antigen Noxa
Clone 69N15C9
Concentration 0.5 mg/mL
Dilution Western Blot 5 - 10 ug/mL
Applications Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP225251SS
SDP
View Documents
Novus Biologicals
NBP225251SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NBP225251
SDP
View Documents
Novus Biologicals
NBP225251
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

Noxa Monoclonal specifically detects Noxa in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Noxa
0.5 mg/mL
Western Blot
Unconjugated
Rat
Apoptosis, Cancer, Cellular Markers, Membrane Trafficking and Chaperones, Plasma Membrane Markers, Stem Cell Markers, Tumor Suppressors
NP_067426.1
5366
This Noxa antibody (69N15C9) is made against amino acids 1-103 of mouse Noxa.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
11.6 kDa
69N15C9
Western Blot 5 - 10 ug/mL
Monoclonal
Purified
RUO
Mouse
adult T cell leukemia-derived PMA-responsive, APR, Immediate-early-response protein APR, NOXAPMA-induced protein 1, phorbol-12-myristate-13-acetate-induced protein 1, PMA-Induced Protein 1, Protein Noxa
PMAIP1
IgG2b κ
Protein G purified
This product reacts with Mouse.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.