Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NuPAGE™ LDS Sample Buffer (4X)

Catalog No. np0007
Encompass_Preferred
Click to view available options
Quantity:
10mL
250mL

Includes

NuPAGE LDS Sample Buffer (4X), 10mL

Used to prepare protein samples for denaturing gel electrophoresis with NuPAGE™ Novex™ gels. Invitrogen™ Novex™ NuPAGE™ LDS Sample Buffer (4X) contains lithium dodecyl sulfate, pH 8.4, which allows for maximum activity of the reducing agent.

Optimal protein sample preparation for polyacrylamide gel electrophoresis (PAGE) requires denaturing and reducing protein disulfide bonds.

  • Highly viscous and concentrated solution that contains higher a concentration of glycerol
  • Contains twice the amount of LDS compared to the amount of SDS in Novex™ Tris-Glycine SDS Sample Buffer or in Tricine SDS Sample Buffer
  • NuPAGE™ LDS Sample Buffer should be brought to room temperature (25°C) prior to use

Order Info

Shipping Condition: Approved for shipment at Room Temperature or on Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Sample Buffer (4X) containing lithium dodecyl sulfate (LDS) at pH 8.5 with SERVA Blue G250 and phenol red.

Store at 4°C to 25°C.

Buffer Sample Loading Buffers
Concentration 4 X
Gel Type NuPAGE
Product Type LDS Sample Buffer
Gel Compatibility NuPAGE™ Gels
Product Line NuPAGE™
Quantity 10mL
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.