Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Npas4 Antibody (S408-79), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP259332R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259332R 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259332R Supplier Novus Biologicals Supplier No. NBP259332R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Npas4 Monoclonal specifically detects Npas4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen Npas4
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone S408-79
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BHLHE79, bHLHe79neuronal PAS domain-containing protein 4, Class E basic helix-loop-helix protein 79, Le-PAS, neuronal PAS domain protein 4, Neuronal PAS4, NXFHLH-PAS transcription factor NXF, PASD10PAS domain-containing protein 10
Gene Symbols NPAS4
Host Species Mouse
Immunogen Fusion protein amino acids 597-802 (C-terminus) of rat Npas4.
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 266743
Test Specificity Detects 90kDa.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.