Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPDC-1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP169583 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP169583 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP169583 Supplier Novus Biologicals Supplier No. NBP169583
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NPDC-1 Polyclonal specifically detects NPDC-1 in Human samples. It is validated for Western Blot.

Specifications

Antigen NPDC-1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Alias CAB-, CAB-1, CAB1CAB, DKFZp586J0523, neural proliferation differentiation and control protein 1, neural proliferation, differentiation and control, 1, NPDC-1
Gene Symbols NPDC1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to NPDC1(neural proliferation, differentiation and control, 1) The peptide sequence was selected from the N terminal of NPDC1. Peptide sequence MATPLPPPSPRHLRLLRLLLSGLVLGAALRGAAAGHPDVAACPGSLDCAL.
Molecular Weight of Antigen 34 kDa
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56654
Test Specificity Yeast: 83%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Yeast
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.