Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPIP Antibody, Novus Biologicals™
SDP

Catalog No. NBP156813 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP156813 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP156813 Supplier Novus Biologicals Supplier No. NBP156813

Rabbit Polyclonal Antibody

NPIP Polyclonal specifically detects NPIP in Human samples. It is validated for Western Blot.

Specifications

Antigen NPIP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q86VD5
Gene Alias FLJ42525, FLJ44848, nuclear pore complex interacting protein, nuclear pore complex-interacting protein
Gene Symbols NPIP
Host Species Rabbit
Immunogen Synthetic peptides corresponding to LOC339047(hypothetical protein LOC339047) The peptide sequence was selected from the middle region of LOC339047. Peptide sequence ECLLTPLPPSALPSADDNLKTPAECLLYPLPPSADDNLKTPPECLLTPLP.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9284
Test Specificity Expected identity based on immunogen sequence: Human: 100%;.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.