Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPL Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB169353 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB169353 25 μg
NB169352 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB169353 Supplier Novus Biologicals Supplier No. NBP31713825UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NPL Polyclonal antibody specifically detects NPL in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen NPL
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias C112, C1orf13MGC61869, chromosome 1 open reading frame 13, DHDPS1, dihydrodipicolinate synthetase homolog 1, EC 4.1.3.3, MGC149582, N-acetylneuraminate lyase, N-acetylneuraminate pyruvate lyase (dihydrodipicolinate synthase), N-acetylneuraminate pyruvate-lyase, N-acetylneuraminic acid aldolase, NAL, NALase, NPL1, Sialate lyase, Sialate-pyruvate lyase, Sialic acid aldolase, Sialic acid lyase
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: FVNGTTGEGLSLSVSERRQVAEEWVTKGKDKLDQVIIHVGALSLKESQELAQHAAEIGAD
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 80896
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.