Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPM1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB11061646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11061646 0.1 mL
NB11061646T 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11061646 Supplier Novus Biologicals Supplier No. NB11061646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

NPM1 Polyclonal specifically detects NPM1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen NPM1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Flow Cytometry 1:100, Immunohistochemistry 1:50 - 1:100, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunoprecipitation 1:50, Immunohistochemistry-Paraffin 1:50 - 1:100
Formulation PBS (pH 7.4) with 0.02% Sodium Azide
Gene Alias B23, MGC104254, NPMnucleophosmin/nucleoplasmin family, member 1, Nucleolar phosphoprotein B23, Nucleolar protein NO38, nucleophosmin, nucleophosmin (nucleolar phosphoprotein B23, numatrin), numatrin
Gene Symbols NPM1
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of human mutant Nucleophosmin (within residues 250-294). [Accession# AAW67752]
Molecular Weight of Antigen 37 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Chromatin Research, DNA replication Transcription Translation and Splicing, Mitotic Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4869
Test Specificity This antibody is specific for the mutant form of Nucleophosmin.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.