Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPM1 Rabbit anti-Human, DyLight 488, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB082804 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB082804 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB082804 Supplier Novus Biologicals Supplier No. NBP298708G

Rabbit Polyclonal Antibody

NPM1 Polyclonal specifically detects NPM1 in Human samples. It is validated for ELISA.

Specifications

Antigen NPM1
Applications ELISA
Classification Polyclonal
Conjugate DyLight 488
Formulation 50 mM Sodium Borate
Gene Alias B23, MGC104254, NPMnucleophosmin/nucleoplasmin family, member 1, Nucleolar phosphoprotein B23, Nucleolar protein NO38, nucleophosmin, nucleophosmin (nucleolar phosphoprotein B23, numatrin), numatrin
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant NPM1 (P06748-1; Met9-Leu158).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, Chromatin Research, DNA replication Transcription Translation and Splicing, Mitotic Regulators, Phospho Specific, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4869
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.