Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPRA/NPR1 Antibody (OTI7H7), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB302243 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302243 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302243 Supplier Novus Biologicals Supplier No. NBP273044APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NPRA/NPR1 Monoclonal antibody specifically detects NPRA/NPR1 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen NPRA/NPR1
Applications Western Blot
Classification Monoclonal
Clone OTI7H7
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias ANPa, ANP-A, ANPR-A, ANPRAEC 4.6.1.2, atrial natriuretic peptide receptor 1, Atrial natriuretic peptide receptor type A, atrionatriuretic peptide receptor A, GC-A, Guanylate cyclase A, GUC2A, GUCY2A, natriuretic peptide A type receptor, natriuretic peptide receptor A, natriuretic peptide receptor A/guanylate cyclase A (atrionatriuretic peptidereceptor A), NPRA, NPR-A
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 746-1006 of human NPR1 (NP_000897) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 4881
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.