Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPRA/NPR1 Antibody (OTI7H7), HRP, Novus Biologicals™
SDP

Catalog No. NBP273044H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273044H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273044H Supplier Novus Biologicals Supplier No. NBP273044H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NPRA/NPR1 Monoclonal specifically detects NPRA/NPR1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen NPRA/NPR1
Applications Western Blot
Classification Monoclonal
Clone OTI7H7
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias ANPa, ANP-A, ANPR-A, ANPRAEC 4.6.1.2, atrial natriuretic peptide receptor 1, Atrial natriuretic peptide receptor type A, atrionatriuretic peptide receptor A, GC-A, Guanylate cyclase A, GUC2A, GUCY2A, natriuretic peptide A type receptor, natriuretic peptide receptor A, natriuretic peptide receptor A/guanylate cyclase A (atrionatriuretic peptidereceptor A), NPRA, NPR-A
Gene Symbols NPR1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 746-1006 of human NPRA/NPR1 (NP_000897) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 4881.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.