Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPRB/NPR2 Antibody (1060722), FITC, Novus Biologicals™
SDP

Catalog No. NB302351 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302351 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302351 Supplier Novus Biologicals Supplier No. FAB9725F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NPRB/NPR2 Monoclonal specifically detects NPRB/NPR2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen NPRB/NPR2
Applications Flow Cytometry
Classification Monoclonal
Clone 1060722
Conjugate FITC
Dilution Flow Cytometry
Formulation PBS
Gene Alias AMD1,AMDM,ANPb,ANPRB,Atrial natriuretic peptide receptor 2,ECDM,GCB,GC-B,GUC2B,GUCY2B,NPRB,NPRBi,SNSK
Host Species Mouse
Immunogen Mouse myeloma cell line, NS0-derived human NPRB/NPR2, Arg23-Ile458, Accession # P20594
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, GPCR, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4882
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.