Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ NPS 2143 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Description
NPS 2143 hydrochloride is a selective Ca2+-sensing receptor antagonist. Blocks increases in cytoplasmic Ca2+ concentrations elicited by human Ca2+ receptors expressed in HEK293 cells (IC50 = 43 nM). Also stimulates parathyroid hormone secretion from bovine parathyroid cells (EC50 = 41 nM). Orally active calcilytic compound.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | NPS 2143 hydrochloride |
| Purity | >99% |
| Molecular Weight (g/mol) | 445.38 |
| Inhibitors | NPS 2143 hydrochloride |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction