Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NPS 2143 hydrochloride
GREENER_CHOICE

Catalog No. 362650R
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Catalog No. 36-265-0R

Supplier: Tocris Bioscience™ 3626/50

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective calcium-sensing receptor (CaSR) antagonist; orally active calcilytic agent

NPS 2143 hydrochloride is a selective Ca2+-sensing receptor antagonist. Blocks increases in cytoplasmic Ca2+ concentrations elicited by human Ca2+ receptors expressed in HEK293 cells (IC50 = 43 nM). Also stimulates parathyroid hormone secretion from bovine parathyroid cells (EC50 = 41 nM). Orally active calcilytic compound.

Specifications

Quantity 50 mg
Product Type NPS 2143 hydrochloride
Purity >99%
Molecular Weight (g/mol) 445.38
Inhibitors NPS 2143 hydrochloride

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.