Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NPY Y2 Receptor Rabbit anti-Mouse, Rat, Polyclonal, Neuromics™
Rabbit Polyclonal Antibody
$253.05
Specifications
Antigen | NPY Y2 Receptor |
---|---|
Concentration | 1.14 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-210-0038
![]() |
Neuromics, Inc
RA1411250UL |
50 μL |
Each of 1 for $253.05
|
|
|||||
Description
NPY Y2 Receptor Polyclonal antibody specifically detects NPY Y2 Receptor in Mouse, Rat samples. It is validated for Immunocytochemistry, Immunofluorescence, ImmunohistochemistrySpecifications
NPY Y2 Receptor | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Rat | |
P97295 | |
18167, 66024 | |
IgG | |
Affinity chromatography | |
Antibody |
1.14 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% sodium azide | |
MGC124241; MGC124242; MGC124244; Neuropeptide Y receptor type 2; neuropeptide Y receptor Y2; neuropeptide Y receptor Y2-like; neuropeptide Y/peptide YY-Y2 receptor; Neuropeptide y-y2; neuropeptide Y-Y2 receptor; Npy2r; NPY2-R; NPY-Y2 receptor; Y2 receptor | |
(C)TDSFSEATNV. Cys added for conjugation to KLH | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
NPY2R |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title