Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPY1R Antibody (556153), FITC, Novus Biologicals™
SDP

Catalog No. NB376612 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB376612 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB376612 Supplier Novus Biologicals Supplier No. FAB6400F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NPY1R Monoclonal specifically detects NPY1R in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen NPY1R
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 556153
Conjugate FITC
Dilution Flow Cytometry, CyTOF-reported
Formulation PBS
Gene Alias neuropeptide Y receptor type 1, neuropeptide Y receptor Y1, NPY1-R, NPYR, NPYY1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human NPY1R, Met1-Ile384, Accession # P25929
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 4886
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.