Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NQO-1 Antibody (A180), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB200209V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB200209V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB200209V Supplier Novus Biologicals Supplier No. NB200209V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NQO-1 Monoclonal specifically detects NQO-1 in Human, Mouse, Rat, Canine, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen NQO-1
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone A180
Conjugate DyLight 405
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Accession No. P15559
Gene Alias Azoreductase, DHQU, DIA4azoreductase, diaphorase (NADH/NADPH) (cytochrome b-5 reductase), dioxin-inducible 1, DTDEC 1.6.5.2, DT-diaphorase, Menadione reductase, NAD(P)H dehydrogenase [quinone] 1, NAD(P)H dehydrogenase, quinone 1, NAD(P)H:menadione oxidoreductase 1, NAD(P)H:Quinone acceptor oxidoreductase type 1, NAD(P)H:quinone oxidoreductase 1, NAD(P)H:quinone oxireductase, NMOR1diaphorase-4, Phylloquinone reductase, QR1NMORI, Quinone reductase 1
Gene Symbols NQO1
Host Species Mouse
Immunogen Full length recombinant NQO1 from human lung [UniProt# P15559]
Molecular Weight of Antigen 31 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1728
Test Specificity Does not cross-react with NQO2.
Target Species Human, Mouse, Rat, Canine, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.