Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NQO-2 Rabbit anti-Human, Mouse, Rat, Clone: 5E0B2, Novus Biologicals™
SDP

Catalog No. NB167659 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167659 100 μg
NB167660 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167659 Supplier Novus Biologicals Supplier No. NBP316284100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

NQO-2 Monoclonal antibody specifically detects NQO-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen NQO-2
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5E0B2
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 1 to 4 μg/mL, Immunohistochemistry-Paraffin 1:1000-1:2500
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias dioxin-inducible-2, EC 1.10.99.2, NAD(P)H dehydrogenase, quinone 2, NAD(P)H menadione oxidoreductase 2, dioxin-inducible, NRH dehydrogenase [quinone] 2, NRH:quinone oxidoreductase 2, Quinone reductase 2, ribosyldihydronicotinamide dehydrogenase, ribosyldihydronicotinamide dehydrogenase [quinone]
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human NQO-2 (P16083).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4835
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.