Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NR4A3/NOR1 Antibody (OTI5C2), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP273052UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273052UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273052UV Supplier Novus Biologicals Supplier No. NBP273052UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NR4A3/NOR1 Monoclonal specifically detects NR4A3/NOR1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen NR4A3/NOR1
Applications Western Blot
Classification Monoclonal
Clone OTI5C2
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CHN, CSMF, MINOR, NOR1, TEC
Gene Symbols NR4A3
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 332-626 of human NR4A3/NOR1 (NP_008912) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 8013.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.