Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NR4A3/NOR1 Antibody (OTI5C2), FITC, Novus Biologicals™
SDP

Catalog No. NB302231 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302231 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302231 Supplier Novus Biologicals Supplier No. NBP273052F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NR4A3/NOR1 Monoclonal antibody specifically detects NR4A3/NOR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen NR4A3/NOR1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5C2
Conjugate FITC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias CHN, CSMF, MINOR, NOR1, TEC
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 332-626 of human NR4A3 (NP_008912) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8013
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.