Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRAGE/MAGED1 Antibody (353124), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB348117 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348117 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348117 Supplier Novus Biologicals Supplier No. FAB3835MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NRAGE/MAGED1 Monoclonal antibody specifically detects NRAGE/MAGED1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen NRAGE/MAGED1
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 353124
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias DLXIN-1, MAGE tumor antigen CCF, melanoma antigen family D, 1, melanoma-associated antigen D1, Neurotrophin receptor-interacting MAGE homolog, NRAGE, NRAGE MAGE-D1 antigen
Host Species Mouse
Immunogen E. coli-derived recombinant human NRAGE, Met1-Glu778, Accession # Q9Y5V3
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 9500
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.