Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRAMP1/SLC11A1 Antibody (910825), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB346111 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346111 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346111 Supplier Novus Biologicals Supplier No. FAB8400MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NRAMP1/SLC11A1 Monoclonal specifically detects NRAMP1/SLC11A1 in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen NRAMP1/SLC11A1
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 910825
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias natural resistance-associated macrophage protein 1, LSH, member 1, NRAMP 1, NRAMP1solute carrier family 11 (sodium/phosphate symporters), member 1, NRAMPnatural resistance-associated macrophage protein 1, solute carrier family 11 (proton-coupled divalent metal ion transporters)
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human NRAMP1/SLC11A1, Met1-Gly550, Accession # NP_000569
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6556
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.