Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRAS Antibody (06), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB328614 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB328614 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB328614 Supplier Novus Biologicals Supplier No. NBP305798MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NRAS Monoclonal antibody specifically detects NRAS in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen NRAS
Applications Western Blot, ELISA
Classification Monoclonal
Clone 06
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias ALPS4, GTPase NRas, HRAS1, neuroblastoma RAS viral (v-ras) oncogene homolog, N-ras, N-ras protein part 4, NRAS1, NS6, Transforming protein N-Ras, v-ras neuroblastoma RAS viral oncogene homolog
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human NRAS (Uniprot#: P01111; Met1-Cys186).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4893
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.