Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRAS Antibody (2G9), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP174044A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP174044A70 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP174044A70 Supplier Novus Biologicals Supplier No. NBP174044AF750

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

NRAS Monoclonal specifically detects NRAS in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen NRAS
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 2G9
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias ALPS4, GTPase NRas, HRAS1, neuroblastoma RAS viral (v-ras) oncogene homolog, N-ras, N-ras protein part 4, NRAS1, NS6, Transforming protein N-Ras, v-ras neuroblastoma RAS viral oncogene homolog
Gene Symbols NRAS
Host Species Mouse
Immunogen Recombinant human NRAS (1-186aa) purified from E. coli
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 4893
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.