Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRAS Antibody (OTI5G7), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP27303A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27303A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27303A70 Supplier Novus Biologicals Supplier No. NBP273053AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NRAS Monoclonal specifically detects NRAS in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen NRAS
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5G7
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias ALPS4, GTPase NRas, HRAS1, neuroblastoma RAS viral (v-ras) oncogene homolog, N-ras, N-ras protein part 4, NRAS1, NS6, Transforming protein N-Ras, v-ras neuroblastoma RAS viral oncogene homolog
Gene Symbols NRAS
Host Species Mouse
Immunogen Full length recombinant protein of human NRAS (NP_002515) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4893.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.