Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRBP1 Antibody (OTI5C3), PE, Novus Biologicals™
SDP

Catalog No. NBP273056PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273056PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273056PE Supplier Novus Biologicals Supplier No. NBP273056PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NRBP1 Monoclonal specifically detects NRBP1 in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Flow Cytometry.

Specifications

Antigen NRBP1
Applications Flow Cytometry
Classification Monoclonal
Clone OTI5C3
Conjugate PE
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias BCON3NRBPMADM, FLJ27109, FLJ35541, MUDPNP, multiple domain putative nuclear protein, myeloid leukemia factor 1 adaptor molecule, nuclear receptor binding protein, nuclear receptor binding protein 1, nuclear receptor-binding protein
Gene Symbols NRBP1
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human NRBP1 expression vector.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 29959.0
Target Species Human, Mouse, Rat, Monkey, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.