Learn More
Description
Specifications
Specifications
| Antigen | NRBP2 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.88mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 |
| Formulation | 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal |
| Gene Accession No. | Q9NSY0 |
| Gene Alias | DKFZp434P086, MGC138699, nuclear receptor binding protein 2, nuclear receptor-binding protein 2, pp9320, Transformation-related gene 16 protein, transformation-related protein 16, TRG16, TRG-16 |
| Gene Symbols | NRBP2 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
