Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NrCAM Antibody (MM0495-8E25), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB361670 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361670 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361670 Supplier Novus Biologicals Supplier No. NBP211826APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NrCAM Monoclonal antibody specifically detects NrCAM in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen NrCAM
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0495-8E25
Conjugate APC
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias bravo, hBravo, KIAA0343Neuronal surface protein Bravo, MGC138845, MGC138846, neuronal cell adhesion molecule, Ng-CAM-related, NgCAM-related cell adhesion molecule, nr-CAM
Host Species Mouse
Immunogen Human recombinant NrCAM
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience, Plasma Membrane Markers
Primary or Secondary Primary
Gene ID (Entrez) 4897
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.