Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NrCAM Antibody (S364-51), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB242191488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB242191488 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB242191488 Supplier Novus Biologicals Supplier No. NBP242191AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NrCAM Monoclonal specifically detects NrCAM in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen NrCAM
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S364-51
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias bravo, hBravo, KIAA0343Neuronal surface protein Bravo, MGC138845, MGC138846, neuronal cell adhesion molecule, Ng-CAM-related, NgCAM-related cell adhesion molecule, nr-CAM
Gene Symbols NRCAM
Host Species Mouse
Immunogen Fusion protein amino acids 30-845 (extracellular domain) of mouse NrCAM. Rat: 96% identity (795/822 amino acids identical). Human: 91% identity (753/822 amino acids identical) ∼50% identity with Neurofascin.
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4897
Test Specificity Approx 50% identity with NeurofascinDetects approx 160kDa.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.