Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nrf1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP17782201 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17782201 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP17782201 Supplier Novus Biologicals Supplier No. NBP1778220.1MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Nrf1 Polyclonal specifically detects Nrf1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Nrf1
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.2mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:3000-1:10000, Immunohistochemistry 2 mg/ml - 5ug/ml, Immunohistochemistry-Paraffin 2 mg/ml - 5μg/ml
Formulation 0.02M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. Q9WU00
Gene Alias Alpha palindromic-binding protein, ALPHA-PAL, EWG, NRF-1, nuclear respiratory factor 1
Gene Symbols NRF1
Host Species Rabbit
Immunogen This protein A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a purified recombinant mouse Nrf1 protein corresponding to aa 1- 534 of the native protein.
Purification Method Protein A purified
Quantity 0.1 mg
Primary or Secondary Primary
Gene ID (Entrez) 4899
Test Specificity This protein A purified antibody is directed against mouse NRF1. BLAST analysis was used to suggest reactivity with this protein from mouse, human, chimpanzee, dog.
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.