Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nrf2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP132822 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP132822 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP132822 Supplier Novus Biologicals Supplier No. NBP132822
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Nrf2 Polyclonal antibody specifically detects Nrf2 in Human, Mouse, Rat, Alligator, Avian, Plant, Zebrafish samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Knockdown.

Specifications

Antigen Nrf2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Simple Western reported by internal validation, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:1000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Knockdown Validated
Formulation PBS (pH7), 20% Glycerol with 0.025% Proclin 300
Gene Accession No. Q16236
Gene Alias HEBP1, NF-E2-related factor 2, NFE2-related factor 2, NRF2 erythroid derived 2, like 2, Nrf2 ihc, Nrf2 immunofluorescence, Nrf2 immunohistochemistry, Nrf2 polyclonal, Nrf2 rat, Nrf2 western blot, nuclear factor (erythroid-derived 2)-like 2, nuclear factor erythroid 2-related factor 2, nuclear factor erythroid-derived 2-like 2
Gene Symbols NFE2L2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NRF2. The exact sequence is proprietary.
Molecular Weight of Antigen 68 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 4780
Test Specificity Predicted cross-reactivity based on sequence identity: Cow (85%).
Target Species Human, Mouse, Rat, Alligator, Avian, Bovine, C. elegans, Plant
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.