Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nrf2, p Ser40 Antibody (SU0334), Novus Biologicals™
SDP

Catalog No. NBP267465 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267465 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267465 Supplier Novus Biologicals Supplier No. NBP267465
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody has been used in 2 publications

Nrf2 Monoclonal specifically detects Nrf2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Nrf2 (p Ser40)
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone SU0334
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias HEBP1, NF-E2-related factor 2, NFE2-related factor 2, NRF2 erythroid derived 2, like 2, nuclear factor (erythroid-derived 2)-like 2, nuclear factor erythroid 2-related factor 2, nuclear factor erythroid-derived 2-like 2
Gene Symbols NFE2L2
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to residues surrounding Ser40 of Human Nrf2 aa 20-69/ 605. (SwissProt: Q16236 Human)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Phospho Specific, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 4780
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.