Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRG4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB186691 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
200 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB186691 50 μg
NB186689 100 μg
NB186690 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. NB186691 Supplier Novus Biologicals Supplier No. NBP31281150UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NRG4 Polyclonal antibody specifically detects NRG4 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen NRG4
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:100-1:500
Formulation 0.2μm filtered solution in PBS without Preservative
Gene Alias DKFZp779N0541, DKFZp779N1944, heregulin 4, HRG4pro-NRG4, neuregulin 4, pro-neuregulin-4, membrane-bound isoform, Pro-NRG4
Host Species Rabbit
Immunogen Purified, recombinant Human NRG4 (Q8WWG1; Pro2-Phe62)
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 145957
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.